Digi-Sense RTD 3 Wire with Transmitter & Thrm
Representative image only.
Digi-Sense RTD 3 Wire with Transmitter & Thrm
Digi-Sense | Item: 3900020
Call For Price
for an additional
₹29,100.00INR / Each
for an additional
₹39,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Digi-Sense RTD 3 Wire with Transmitter & Thrm
Digi-Sense RTD 3 Wire with Transmitter & Thrm
3900020
Call For Price
+ =
Total Price 39900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 316 Stainless steel sheath material
  • 1/2'' X 1/2'' NPT stainless steel hex nipple for head mounting fittings
  • Aluminum screw cover connection head
  • Low range or high range wire wound elements
  • .00385º C temperature coefficient
  • Custom built assemblies available upon request
  • 1 year warranty
More About this Item
These RTD assemblies consist of a single Low Range or High Range Wire Wound element constructed of 316 Stainless Steel sheath material, assembled together with an Aluminum screw cover connection head.