Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, PP; 10/pk
Representative image only.
Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, PP; 10/pk
Diba | Mfr: 002301| Item: 2193996
Solution for quick and easy connection
₹25,300.00INR / Pkg of 10
₹25,300.00INR / Pkg of 10
Buy It Together
Current Item
Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, PP; 10/pk
Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, PP; 10/pk
2193996
₹25,300.00INR / Pkg of 10
+ =
Total Price 30200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Available in polypropylene (PP), ETFE, or PEEK materials
More About this Item
Easily connect 1/4"-28 UNF(M) fittings with compatible unions.
Specifications
  • Length (")3/4
  • Fitting MaterialPolypropylene
  • Width (InchShort)23/64
  • Connection TypeUNF(F) x UNF(F)
  • Number Of Ports2
  • Thread size UNF(F) (port 2) (InchShort)1/4"-28
  • Thread size UNF(F) (port 1) (InchShort)1/4"-28
  • ColorWhite
  • Orifice Dimensions (mm)1.5
  • DescriptionDibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, PP; 10/pk