Dibafit™ Blanking Plug, 1/4"-28 UNF(M) flat bottom, ETFE; 2/pk
Representative image only.
Dibafit™ Blanking Plug, 1/4"-28 UNF(M) flat bottom, ETFE; 2/pk
Diba | Mfr: 002320| Item: 2194004
Optimize performance of your Diba Omnifit® products
₹4,900.00INR / Pkg of 2
₹4,900.00INR / Pkg of 2
Buy It Together
Current Item
Dibafit™ Blanking Plug, 1/4"-28 UNF(M) flat bottom, ETFE; 2/pk
Dibafit™ Blanking Plug, 1/4"-28 UNF(M) flat bottom, ETFE; 2/pk
2194004
₹4,900.00INR / Pkg of 2
+ =
Total Price 19900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quality and consistency your lab can count on
More About this Item
Choose from various fittings, filters and traps to meet your application needs.
Specifications
  • MaterialETFE
  • Compatible WithDiba fittings and valves with 1/4"-28 UNF(F) ports
  • Connections1/4"-28 UNF(M)