Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, ETFE; 10/pk
Representative image only.
Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, ETFE; 10/pk
Diba | Mfr: 002302| Item: 2193997
Solution for quick and easy connection
₹7,000.00INR / Pkg of 10
₹7,000.00INR / Pkg of 10
Buy It Together
Current Item
Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, ETFE; 10/pk
Dibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, ETFE; 10/pk
2193997
₹7,000.00INR / Pkg of 10
+ =
Total Price 11900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Available in polypropylene (PP), ETFE, or PEEK materials
More About this Item
Easily connect 1/4"-28 UNF(M) fittings with compatible unions.
Specifications
  • Fitting MaterialETFE
  • Length (")3/4
  • Width (InchShort)23/64
  • Connection TypeUNF(F) x UNF(F)
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)1/4"-28
  • Thread size UNF(F) (port 2) (InchShort)1/4"-28
  • ColorNatural
  • Orifice Dimensions (mm)1.5
  • DescriptionDibafit™ Threaded Coupling, 1/4"-28 UNF(F) flat bottom, ETFE; 10/pk