Dibafit™ Gripper Fitting, M6 ISO thread, 1/16" OD tubing, PEEK; 10/pk
Representative image only.
Dibafit™ Gripper Fitting, M6 ISO thread, 1/16" OD tubing, PEEK; 10/pk
Diba | Mfr: 002135| Item: 2193975
One-piece grippers withstand pressures up to 1000 psi!
₹9,300.00INR / Pkg of 10
₹9,300.00INR / Pkg of 10
Buy It Together
Current Item
Dibafit™ Gripper Fitting, M6 ISO thread, 1/16" OD tubing, PEEK; 10/pk
Dibafit™ Gripper Fitting, M6 ISO thread, 1/16" OD tubing, PEEK; 10/pk
2193975
₹9,300.00INR / Pkg of 10
+ =
Total Price 28200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Permanently attached PTFE ferrules are ideal for repeated connect/disconnect
  • Inner PTFE sealing face is the only material in the fluid path
More About this Item
Dibafit gripper fitting system uses a one-piece ferrule that permanently attaches to the outside of semi-rigid tubing. The fitting can spin freely, preventing tubing from twisting. PEEK fittings withstand higher temperatures and provide excellent chemical resistance.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeCompression x ISO(M)
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.6
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorNatural
  • DescriptionGripper Fitting, M6 ISO thread, 1/16" OD tubing, PEEK; 10/pk