Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, M6 ISO Thread x 1/16" OD; 10/Pk
Representative image only.
Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, M6 ISO Thread x 1/16" OD; 10/Pk
Diba | Mfr: 002313| Item: 2194002
Chemically inert flangeless fitting with permanently attached PTFE ferrule ideal for repeat connect/disconnect
₹11,100.00INR / Pkg of 10
₹11,100.00INR / Pkg of 10
Product Description
Key Features
  • Available in PTFE/SS or PTFE/PEEK material
More About this Item
The Dibafit™ Gripper fitting system uses a one-piece ferrule which fits permanently onto the outside of semi-rigid tubing.