Diba Omnifit® Standard Flow Bubble Trap, PEEK™, 10 µm PTFE filter; 1/ea
Representative image only.
Diba Omnifit® Standard Flow Bubble Trap, PEEK™, 10 µm PTFE filter; 1/ea
Diba | Mfr: 006BT| Item: 2194038
Remove bubbles with or without vacuum assistance
₹35,000.00INR / Each
₹35,000.00INR / Each
Buy It Together
Current Item
Diba Omnifit® Standard Flow Bubble Trap, PEEK™, 10 µm PTFE filter; 1/ea
Diba Omnifit® Standard Flow Bubble Trap, PEEK™, 10 µm PTFE filter; 1/ea
2194038
₹35,000.00INR / Each
+ + =
Total Price 50900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed with durable PTFE membrane
More About this Item
Effectively remove bubbles* from aqueous solutions inline or downstream with bubble traps. Traps feature ¼"-28 UNF(F) port and come with 10 µm PTFE filter. The high-flow trap features a vacuum port on the atmosphere (dry) side of the membrane. When vacuum is applied, the pressure differential between the system and atmosphere side increases (must not exceed 30 psi/2.1 bar), causing bubbles to be removed from the liquid and permitting increased liquid flow.
*These bubble traps do not remove dissolved gases.
Specifications
  • MaterialPEEK™ body, PTFE membrane
  • Flow Rate6 mL/min (max)
  • Port Size1/4"-28 UNF(F)
  • Max Pressure (PSI)30