Diba Omnifit® Manual Sample Injection Valve; 1/ea
Representative image only.
Diba Omnifit® Manual Sample Injection Valve; 1/ea
Diba | Mfr: 1106| Item: 2193918
Designed for low-pressure chromatography systems
₹156,000.00INR / Each
₹156,000.00INR / Each
Buy It Together
Current Item
Diba Omnifit® Manual Sample Injection Valve; 1/ea
Diba Omnifit® Manual Sample Injection Valve; 1/ea
2193918
₹156,000.00INR / Each
+ =
Total Price 160900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Choose from single or 6-port valve
More About this Item
Manual sample injection valve 21939-18 is designed for low-pressure chromatography systems. Easily introduce reproducible sample volumes using the included 0.5 mm, 1.0 mm, and 2.5 mm loops and 5-mL syringe.
Six-port loop injection valve 21940-24 has a "click-stop" mechanism for easy port identification. It is constructed from PTFE with a PEEK™ casing and PCTFE rotor and withstands pressures up to 500 psi (33 bar). Valve accepts 1/4"-28 UNF(M) fittings and features a 0.8 mm bore and M3 mounting holes. Use plug 21940-04 to seal any unused ports.
Specifications
  • Connection Type1/4"-28 UNF(F)
  • Inlet Port1 port
  • Max Pressure (PSI)300
  • Max Pressure (bar)20.5