Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk
Representative image only.
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk
Diba | Mfr: 008NC47-YC7U| Item: 2194192
Removable and reuable system provides quick and convenient connections
₹7,000.00INR / Pkg of 10
₹7,000.00INR / Pkg of 10
Buy It Together
Current Item
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk
2194192
₹7,000.00INR / Pkg of 10
+ =
Total Price 19600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • ETFE cone is the only material in the fluid path, providing excellent chemical resistance
  • Compact head fittings are great for tight spaces
More About this Item
None of the parts are permanently attached to the tubing, so both fittings and inverted cones can easily be removed and reused. Select from short or long inverted cones (ferrules). Short inverted cones are designed to give most thread engagement in shallow ports.
Specifications
  • Fitting MaterialPolypropylene
  • Length (")15/16
  • Width (InchShort)5/16
  • Connection TypeCompression x UNF(M)
  • Number Of Ports2
  • Thread size UNF(M) (port 1) (InchShort)5/16"-24
  • Compression Size / Tube Od (Port 2) (in)3/16
  • ColorBlue
  • Max Operating Pressure (PSI)200
  • DescriptionOmni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk