DeFelsko PosiTector 200B1 Coating Thickness Gauge, Non-Metals, 0.5-40 mils
Representative image only.
DeFelsko PosiTector 200B1 Coating Thickness Gauge, Non-Metals, 0.5-40 mils
DeFelsko | Mfr: 200B1| Item: 5978402
Quickly measure coating thickness on all non-metal substrates
₹582,000.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹582,000.00INR / Each
Buy It Together
Current Item
DeFelsko PosiTector 200B1 Coating Thickness Gauge, Non-Metals, 0.5-40 mils
DeFelsko PosiTector 200B1 Coating Thickness Gauge, Non-Metals, 0.5-40 mils
5978402
₹582,000.00INR / Each
+ +
DeFelsko GEL P Ultrasonic Couplant, Gel; 12/Cs
5978429
₹58,300.00INR / Each
=
Total Price 677700.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Ready to measure - no calibration adjustment required to measure most coatings
  • Solvent, acid, oil, water and dust resistant - meets or exceeds IP5X
  • Internal memory for 250 readings in a single batch to be view or downloaded
  • Statistics mode continuously displays average, standard deviation, and number of readings while measuring
  • High contrast backlit display for bright or dark environments
  • Certificate of calibration showing traceability to NIST or PTB included (Long Form)
  • Proven non-destructive ultrasonic technique conforms to ASTM6132 and ISO2808
  • Selectable display languages
  • Non-destructively measures thickness of coating 0.5 to 40 mils (13 to 100 microns)
  • 2 year warranty
More About this Item
The standard ultrasonic coating thickness gauges non-destructively measure the thickness of coatings on non-metal substrates including wood, plastic, composites, and more. They also measure the total thickness of a coating system.
Two year warranty
Specifications
  • Range0.5 to 40 mil
  • DisplayLCD
  • DescriptionCoating Thickness Gauge, Non-Metals, 0.5-40 mils