DeFelsko GEL P Ultrasonic Couplant, Gel; 12/Cs
Representative image only.
DeFelsko GEL P Ultrasonic Couplant, Gel; 12/Cs
DeFelsko | Mfr: GEL P| Item: 5978429
Ensure accurate thickness measurements with ultrasonic gel
₹58,300.00INR / Each
₹58,300.00INR / Each
Product Description
Key Features
  • Compatible with DeFelsko PosiTector® 200 and PosiTector® UTG ultrasonic thickness gauges
  • 12 x 4 oz bottles
  • 2 year warranty
More About this Item
Gel acts as a medium between the transducer and the sample to help produce accurate and reliable readings.
Two year warranty
Specifications
  • Compatible WithCompatible with DeFelsko PosiTector® 200 59784-01 and 59784-02 and PosiTector® UTG 59784-52 and 59785-12 ultrasonic thickness gauges
  • DescriptionUltrasonic Couplant, Gel; 12/Cs