Corning FTS30 CoolCell Vial Module for 30 Vials; 10/pk
Representative image only.
Corning FTS30 CoolCell Vial Module for 30 Vials; 10/pk
Corning | Mfr: 432078| Item: 0439278
Increase efficiency and throughput
₹43,800.00INR / Case of 10
₹43,800.00INR / Case of 10
Buy It Together
Current Item
Corning FTS30 CoolCell Vial Module for 30 Vials; 10/pk
Corning FTS30 CoolCell Vial Module for 30 Vials; 10/pk
0439278
₹43,800.00INR / Case of 10
+
Corning Cryoceps Vial Grippers, 5pk
0439336
₹20,300.00INR / Each
=
Total Price 64100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Use extra modules to quickly remove frozen samples from the CoolBox container, then load container with fresh samples. Modules are compatible with standard 5" x 5" x 2" (12.7 x 12.7 x 5.1 cm) cryogenic storage boxes.
1 Year Warranty
Specifications
  • Capacity (mL)1.0 to 2.0
  • Compatible WithFTS30 CoolCell containers
  • DescriptionVial Module for 30 Vials; 10/pk