Corning CryoCeps Vial Grippers, 5pk
Representative image only.
Corning CryoCeps Vial Grippers, 5pk
Corning | Mfr: 432136| Item: 0439336
Protect your fingers from dry ice or liquid nitrogen burns
₹20,300.00INR / Each
₹20,300.00INR / Each
Product Description
Key Features
  • Designed to easily and securely grip cryovials
  • Replaces forceps—eliminates slipping and dropping of vials
  • 1 year warranty
More About this Item
Cryogenic vial grippers are specially designed to grasp internal or external-thread cryogenic vials, protecting your fingers from contacting frozen vials, dry ice, and liquid nitrogen. Securely sort and move vials while maintaining sterility. Pack contains 5 grippers; one each of green, pink, purple, orange, and blue.
1 Year Warranty
Specifications
  • ColorGreen, pink, purple, orange, and blue
  • Length (in)5 1/2
  • Length (cm)14
  • DescriptionCryogenic vial grippers, polypropylene copolymer