Corning 3005 Single-use Sterile spatula, round end/spoon
Representative image only.
Corning 3005 Single-use Sterile spatula, round end/spoon
Corning | Mfr: 3005| Item: 0626504
Call For Price
Call For Price
Buy It Together
Current Item
Corning 3005 Single-use Sterile spatula, round end/spoon
Corning 3005 Single-use Sterile spatula, round end/spoon
0626504
Call For Price
+ +
Cole-Parmer Spatula Macro Spoon, Stainless Steel,120mm; 3/PK
0628704
₹1,900.00INR / Pkg of 3
+ + =
Total Price 19500.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Modified polystyrene construction—nontoxic and chemical resistant
More About this Item
Never waste time resterilizing spatulas again. Each spatula is individually packaged and comes gamma irradiation sterilized for no ETO residues. Spatulas are RNase/DNase free, nonpyrogenic, and antistatic.
BioPyr
Specifications
  • Overall Length (in)9 11/16
  • Overall Length (cm)24.61
  • MaterialPolystyrene
  • SterileYes
  • DescriptionSingle-use Sterile spatula, round end/spoon