COMARK SK24M Thermocouple Heavy Duty General Purpose Surface Probe, Type K, 10 mm Stem dia
Representative image only.
COMARK SK24M Thermocouple Heavy Duty General Purpose Surface Probe, Type K, 10 mm Stem dia
COMARK | Item: 0887985
Designed specifically for use in the food industry
₹29,700.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹29,700.00INR / Each
Buy It Together
Current Item
COMARK SK24M Thermocouple Heavy Duty General Purpose Surface Probe, Type K, 10 mm Stem dia
COMARK SK24M Thermocouple Heavy Duty General Purpose Surface Probe, Type K, 10 mm Stem dia
0887985
₹29,700.00INR / Each
+ =
Total Price 72400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Most probes have a response time of under 0.5 seconds
  • Include a 7-foot coil cord ( except for the ATT29 probe which is 4-foot cord )
  • 0.4 second response time
  • Temperature range: -58° to +1202°F (-50 to 650°C)
  • Six month warranty
More About this Item
Comark food probes are designed specifically for use in the food industry to ensure proper food preparation. Penetration probes are designed to be inserted directly into the food, typically poultry, beef or pork, to gauge the internal temperature and to ensure that the meat is cooked properly for safe eating. Surface probes are used to make sure the cooking surface is at the desired temperature before and during the cooking process.
6 Month Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-58
  • Max Temperature (° F)1202
  • Min Temperature (° C)-50
  • Max Temperature (° C)650
  • Response Time (Sec)0.4
  • Connector TypeSub-mini (2-pin)
  • DescriptionProbe, Surface Hvy Duty Type K 4" Heavy Duty 3 Second Response