Cole-Parmer Vortex Mixer with Accessory Pack
Representative image only.
Cole-Parmer Vortex Mixer with Accessory Pack
Cole-Parmer | Item: 0472905
Compact unit perfect for individual work stations
₹64,300.00INR / Each
₹64,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer Vortex Mixer with Accessory Pack
Cole-Parmer Vortex Mixer with Accessory Pack
0472905
₹64,300.00INR / Each
+ + +
Cole-Parmer Vortex Mixer/Shaker Accessory Cup Pack
0472907
₹10,500.00INR / Each
+
Cole-Parmer Variable Speed Vortex Mixer; 90 to 240 VAC
0475051
₹57,800.00INR / Each
=
Total Price 225000.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Standard rubber cup and accessory pack: plastic cradle for standard microplate, circular foam insert for beakers and flasks (up to 500 mL capacity), rectangular foam insert (holds 8 x 0.2 mL, 8 x 0.5 mL and 16 x 1.5 mL microcentrifuge tubes), and solid rectangular foam insert (blank for custom holes).
Key Features
  • Mix a wide variety of tube sizes and shapes with the included rubber cup head
  • Heavy die-cast body won't dance across your bench
  • Use for continuous mixing — housing design supports use with a lab rod
  • Accessory pack expands mixing capabilities
  • 3 year warranty
More About this Item
Vortex mixer features a robust die-cast body to prevent traveling while in use, and has an integrated port to support a lab rod for holding vessels in place during continuous operation.
3 Year Warranty
Specifications
  • DescriptionVortex Mixer with FREE Accessory Pack
  • Warranty3 Years