Cole-Parmer Venturi Miniature Vacuum Pump, 0.5 cfm, 27.0"Hg

Representative image only.

Cole-Parmer Venturi Miniature Vacuum Pump, 0.5 cfm, 27.0"Hg

Cole-Parmer | Item: 7816540

No moving parts ensures long life with low maintenance

₹20,500.00INR / Each

₹20,500.00INR / Each

Buy It Together

Current Item

Cole-Parmer Venturi Miniature Vacuum Pump, 0.5 cfm, 27.0"Hg
Cole-Parmer Venturi Miniature Vacuum Pump, 0.5 cfm, 27.0"Hg
7816540

₹20,500.00INR / Each
+ + + =
Total Price 436900.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Venturi pumps are air-powered—require no electrical power source or lubricant
  • Place these compact vacuum pumps close to application for fast response and minimal vacuum loss
  • 5 year warranty

More About this Item

These pumps take advantage of the venturi effect—as a stream of compressed air exits a constricted orifice the resulting pressure drop draws air through a vacuum port. Since they require no oil or electricity, the resulting vacuum (up to 29.5" Hg) is clean and efficient. With no moving parts or electrical hazards, these air-powered pumps are also intrinsically safe. The remarkably compact size of these pumps enables you to design vacuum systems with smaller, less expensive components and reduced maintenance costs.

Select the model of vacuum pump best suited to your specific application. Model 78165-20 includes a vacuum gauge which provides easy visual monitoring—helpful when troubleshooting. We recommend an air filter upstream of the pump to prolong service life. Filter should have a pore size of 50 µm or smaller.

5 Year Warranty
Specifications
  • Wetted PartsAluminum
  • Free-Air Capacity (CFM)0.3
  • Free-Air Capacity (LPM)8.5
  • Free-Air Capacity (LPH)510
  • Max Flow Rate (CFM)0.3
  • Max Flow Rate (Liters/min)8.5
  • Max Vacuum (in Hg)27
  • Air Input At 80 Psi (CFM)0.75
  • Air Input At 80 Psi (LPM)21.2
  • Port Size10-32 UNF
  • Length (in)1.25
  • Diameter (in)0.5
  • Length (cm)3.18
  • Diameter (cm)1.27
  • Length (mm)31.8
  • Diameter (mm)12.7
  • DescriptionVenturi miniature vacuum pump, 0.5 cfm, 27.0
  • Warranty5 Years
Reviews
Q&A