CPS Single and Double Stage Vacuum Pumps
The PRO-SET® range of dual voltage vacuum pumps include both single and two stage pump configurations ranging in size from 2 to 12 CFM. All units incorporate heavy-duty dual voltage motors that can operate on either 115 VAC or 230 VAC (50 or 60 Hz) power at the flick of a switch. Steel
reinforced carrying handles are over molded for comfort and proprietary hard rubber bases absorb vibration for smoother, quieter operation. All units incorporate integrated gas ballast valves, multi-threaded connection ports and are overload protected.
45,200.00 - ₹186,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (CFM) Max Vacuum (micron)
Availability Pricing
CPS Products VP10D Pro-Set Two Stage Vacuum Pump, 10 cfm, 10 micron 0716481 Mfr # VP10D
COMPARE
10 10
₹169,000.00INR / Each
CPS Products VP12D Pro-Set Two Stage Vacuum Pump, 12 cfm, 10 micron 0716482 Mfr # VP12D
COMPARE
12 10
₹186,000.00INR / Each
CPS Products VP2D Pro-Set Two Stage Vacuum Pump, 2 cfm, 10 micron 0716483 Mfr # VP2D
COMPARE
2 10
₹69,800.00INR / Each
CPS Products VP2S Pro-Set Single Stage Vacuum Pump, 2 cfm, 10 micron 0716484 Mfr # VP2S
COMPARE
2 10
₹45,200.00INR / Each
CPS Products VP4S Pro-Set Single Stage Vacuum Pump, 3 cfm, 10 micron 0716486 Mfr # VP4S
COMPARE
3 10
₹56,400.00INR / Each
CPS Products VP6D Pro-Set Two Stage Vacuum Pump, 6 cfm, 10 micron 0716487 Mfr # VP6D
COMPARE
6 10
₹102,000.00INR / Each
CPS Products VP6S Pro-Set Single Stage Vacuum Pump, 6 cfm, 50 micron 0716488 Mfr # VP6S
COMPARE
6 50
₹70,900.00INR / Each
CPS Products VP8D Pro-Set Two Stage Vacuum Pump, 8 cfm, 10 micron 0716489 Mfr # VP8D
COMPARE
8 10
₹128,000.00INR / Each
Showing 1- 8 of 8