Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, PTFE, 1/4" NPT(M) x 5/16-24 UNF(F)
Representative image only.
Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, PTFE, 1/4" NPT(M) x 5/16-24 UNF(F)
Cole-Parmer | Item: 1202083
Designed for secure leak-resistant connections
₹5,600.00INR / Each
₹5,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, PTFE, 1/4" NPT(M) x 5/16-24 UNF(F)
Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, PTFE, 1/4" NPT(M) x 5/16-24 UNF(F)
1202083
₹5,600.00INR / Each
+ + + + =
Total Price 28100.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • 9/16" hex body makes it easy to wrench tighten adapters
  • Thread to thread adapters provide a tight seal
  • Constructed of chemically inert PTFE
More About this Item
These Cole-Parmer® Vaplock™ threaded adapters allow for quick and easy connection of different thread sizes. Chromatography adapter fittings are ideal for use in Vaplock manifolds, solvent delivery and waste collection, and general chromatography applications.
Specifications
  • Fitting MaterialPTFE
  • Number Of Ports2
  • Connection Type1/4" NPT(M) x 5/16-24 UNF(F)
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Thread size UNF(F) (port 2) (InchShort)5/16-24
  • Flow PatternStraight
  • ColorWhite
  • DescriptionThreaded Adapter Fitting, 9/16" Hex, Straight, PTFE, 1/4" NPT(M) x 5/16-24 UNF(F)