Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, Polypropylene, 1/4" NPT(M) x 5/16-24 UNF(F)
Representative image only.
Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, Polypropylene, 1/4" NPT(M) x 5/16-24 UNF(F)
Cole-Parmer | Item: 1202079
Designed for secure leak-resistant connections
₹2,300.00INR / Each
₹2,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, Polypropylene, 1/4" NPT(M) x 5/16-24 UNF(F)
Cole-Parmer VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, Polypropylene, 1/4" NPT(M) x 5/16-24 UNF(F)
1202079
₹2,300.00INR / Each
+ + + + =
Total Price 25200.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • 9/16" hex body makes it easy to wrench tighten adapters
  • Thread to thread adapters provide a tight seal
  • Constructed of durable polypropylene
More About this Item
These Cole-Parmer® Vaplock™ threaded adapters allow for quick and easy connection of different thread sizes. Chromatography adapter fittings are ideal for use in Vaplock manifolds, solvent delivery and waste collection, and general chromatography applications.
Specifications
  • Fitting MaterialPolypropylene
  • Connection Type1/4" NPT(M) x 5/16-24 UNF(F)
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Thread size UNF(F) (port 2) (InchShort)5/16-24
  • Flow PatternStraight
  • ColorTranslucent white
  • DescriptionThreaded Adapter Fitting, 9/16" Hex, Straight, Polypropylene, 1/4" NPT(M) x 5/16-24 UNF(F)