Cole-Parmer VapLock™ Solvent Delivery Cap, four 1/4"-28 ports, 38-430; 1/ea
Representative image only.
Cole-Parmer VapLock™ Solvent Delivery Cap, four 1/4"-28 ports, 38-430; 1/ea
Cole-Parmer | Item: 1201829
Reduce chemical evaporation and exposure to hazardous vapors
₹5,600.00INR / Each
₹5,600.00INR / Each
Product Description
Key Features
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
More About this Item
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.
Caps with threaded port have ports angled at 10° tilt for more accessible tubing connections. Select caps with stainless steel (SS) thread insert to minimize cross-threading and damage to soft PTFE ports.
*Does not come in contact with liquid under normal circumstances.
Specifications
  • Cap MaterialPTFE/PP
  • Cap / Stopper SizeNalgene 38-430
  • Cap ColorWhite
  • Special Features4 x 1/4-28 UNF(F)
  • AutoclavableYes
  • DescriptionSolvent Delivery Cap, four 1/4-28 ports, 38-430