Cole-Parmer VapLock™ Reversible Control Valve, 2 x 1/4" NPT(M), 0.1-0.25 psi crack pressure, PP & EPDM; 1ea
Representative image only.
Cole-Parmer VapLock™ Reversible Control Valve, 2 x 1/4" NPT(M), 0.1-0.25 psi crack pressure, PP & EPDM; 1ea
Cole-Parmer | Item: 1202058
Maintain pressure equilibrium within the bottle for safe solvent handling
₹6,900.00INR / Each
₹6,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Reversible Control Valve, 2 x 1/4" NPT(M), 0.1-0.25 psi crack pressure, PP & EPDM; 1ea
Cole-Parmer VapLock™ Reversible Control Valve, 2 x 1/4" NPT(M), 0.1-0.25 psi crack pressure, PP & EPDM; 1ea
1202058
₹6,900.00INR / Each
+ =
Total Price 13500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Valves feature 0.1 PSI breaking pressure
More About this Item
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Reversible valves serve either purpose depending on the orientation of the valve.
Specifications
  • Connection Type1/4" NPT(M) x 1/4" NPT(M)
  • Breaking Pressure (PSI)0.1
  • Seal MaterialEPDM
  • Housing MaterialPP
  • Wetted MaterialsPP/EPDM
  • DescriptionReversible Valve, 2 x 1/4" NPT(M), 0.1-0.25 psi crack pressure, PP & EPDM; 1ea