Cole-Parmer VapLock™ Relief Valve, 1/4-28 UNF(M), 1-2.5 psi crack pressure, PTFE & Viton; 1ea
Representative image only.
Cole-Parmer VapLock™ Relief Valve, 1/4-28 UNF(M), 1-2.5 psi crack pressure, PTFE & Viton; 1ea
Cole-Parmer | Item: 1202056
Maintain pressure equilibrium within the bottle for safe solvent handling
₹6,600.00INR / Each
₹6,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Relief Valve, 1/4-28 UNF(M), 1-2.5 psi crack pressure, PTFE & Viton; 1ea
Cole-Parmer VapLock™ Relief Valve, 1/4-28 UNF(M), 1-2.5 psi crack pressure, PTFE & Viton; 1ea
1202056
₹6,600.00INR / Each
+ + + =
Total Price 24900.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Choice of 0.1 to 0.25 psi or 1 to 2.5 psi cracking pressure
More About this Item
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Relief valves let pressurized bottle contents escape above the cracking pressure.
Specifications
  • Connection Type1/4"-28 UNF(M)
  • Breaking Pressure (PSI)1
  • Seal MaterialViton®
  • Housing MaterialPTFE
  • Wetted MaterialsPTFE/Viton®
  • DescriptionRelief Valve, 1/4-28 UNF(M), 1-2.5 psi crack pressure, PTFE & Viton; 1ea