Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 5/32" ID x 1/4" NPT(M); 1/EA
Representative image only.
Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 5/32" ID x 1/4" NPT(M); 1/EA
Cole-Parmer | Item: 1202634
Designed for secure leak-resistant connections
₹130.00INR / Each
₹130.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Fitting, Polypropylene,  Straight, Hose Barb to Threaded Adapter, 5/32" ID x 1/4" NPT(M); 1/EA
Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 5/32" ID x 1/4" NPT(M); 1/EA
1202634
₹130.00INR / Each
+ + + + + + + + + =
Total Price 30930.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Constructed of durable polypropylene
More About this Item
Adapters for flexible tubing let you connect 1/16" to 3/4" ID soft-wall tubing to NPT(F) ports. Adapters are made of polypropylene. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeBarb x NPT(M)
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • Barb Size / Tubing Id (Port 1) (in)5/32
  • ColorGray
  • Description Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 5/32" ID x 1/4" NPT(M); 1/EA