Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/CPVC/3kOhm Thermistor; Spade/BNC
Representative image only.
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/CPVC/3kOhm Thermistor; Spade/BNC
Cole-Parmer | Item: 2700311
Less chance of breakage
₹34,900.00INR / Each
₹34,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/CPVC/3kOhm Thermistor; Spade/BNC
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/CPVC/3kOhm Thermistor; Spade/BNC
2700311
₹34,900.00INR / Each
+ =
Total Price 51200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Feature a 10-ft cable with BNC connector
  • 1 year warranty
More About this Item
pH electrodes are ideal for low temperatures, nonaqueous solutions, and pH measurements from 0 to 12.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Min pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)3/4
  • ATC ConnectorSpade lugs
  • ATC Element3 kΩ thermistor
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionTuff-Tip In-Line pH Probe, DJ/CPVC/3kOhm Thermistor; Spade/BNC