Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, White Nylon, Cleanroom Packed, 1/8" NPT(M) x 3/8" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, White Nylon, Cleanroom Packed, 1/8" NPT(M) x 3/8" ID; 10/Pk
Cole-Parmer | Item: 5010777
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹3,200.00INR / Pkg of 10
₹3,200.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Meets FDA regulations 21 CFR 178.3297 and is BSE/TSE and RoHS compliant
  • 1/8" NPT(M) x 3/8" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These nylon fittings are made from the same resin as natural nylon but with FDA-compliant white coloring. They offer good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 121°C
  • Connection TypeThreaded NPT(M) x Hose Barb
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • Flow PatternStraight
  • ColorWhite
  • Regulatory ApprovalsRoHS;Non-DEHP;Non-Phthalate;FDA
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, White Nylon, Cleanroom Packed, 1/8" NPT(M) x 3/8" ID; 10/Pk