Cole-Parmer TELOS® Column Sample Processing Manifold, 12 columns, 16 x 100 mm rack
Representative image only.
Cole-Parmer TELOS® Column Sample Processing Manifold, 12 columns, 16 x 100 mm rack
Cole-Parmer | Item: 0648004
Save time by processing multiple samples simultaneously
₹122,000.00INR / Each
₹122,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer TELOS® Column Sample Processing Manifold, 12 columns, 16 x 100 mm rack
Cole-Parmer TELOS® Column Sample Processing Manifold, 12 columns, 16 x 100 mm rack
0648004
₹122,000.00INR / Each
+ +
Advantec Manifold, Stainless Steel, 1 station
3521155
₹330,000.00INR / Each
=
Total Price 459800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
glass tank, lid, PP needles, stopcocks, rack, vacuum gauge, waste collection container, and plates for accepting 11-, 12-, and 16-mm diameter tubes to a maximum length of 13 cm.
Key Features
  • Glass tank allows a clear view of the extraction process
  • Control sample flow individually using the optional PTFE stopcocks
  • Separate waste container prevents tank contamination
  • Process up to 12 columns at once
More About this Item
Use vacuum manifolds for SPE sample preparation, filtration, and elution. Manifolds come complete with everything you need—tank, lid, needles, stopcocks, rack, vacuum gauge, waste collection container, and plates for accepting different size tubes. Easily control the flow rates by adjusting the vacuum supply to the manifold. Each port can be fitted with a stopcock for individual column control or isolation.
Use the waste container for SPE column processing prior to analyte or compound elution. Having a separate waste container helps keep the main tank clean, preventing potential contamination of the final sample and preventing direct contact with chemical or biohazard samples by the user.
Specifications
  • Branches12
  • DescriptionTELOS Column Sample Processing Manifold, 12 columns, 16 x 100 mm rack