Cole-Parmer Sterile Plastic Pasteur Pipettes, 230 mm, Individually Wrap; 200/PK
Representative image only.
Cole-Parmer Sterile Plastic Pasteur Pipettes, 230 mm, Individually Wrap; 200/PK
Cole-Parmer | Item: 0439512
A safe alternative to glass pipettes
₹10,300.00INR / Pkg of 200
₹10,300.00INR / Pkg of 200
Buy It Together
Current Item
Cole-Parmer Sterile Plastic Pasteur Pipettes, 230 mm, Individually Wrap; 200/PK
Cole-Parmer Sterile Plastic Pasteur Pipettes, 230 mm, Individually Wrap; 200/PK
0439512
₹10,300.00INR / Pkg of 200
+ =
Total Price 14500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Manufactured from medical-grade polystyrene in an ISO Class 8 cleanroom
  • Gamma irradiated, and RNase, DNase, and pyrogen free
More About this Item
Disposable polystyrene (PS) Pasteur pipettes offer the aspirating precision of glass, with the safety and benefits of plastic. Ideal for use with aspirating bulbs or vacuum applications to remove liquid waste from flasks, dishes, plates, and tubes. The long flexible stem can be bent to draw liquid from narrow or small volume tubes. Supplied individually wrapped.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Overall Length (InchShort)9
  • Overall Length (mm)230
  • Body MaterialPolystyrene
  • DescriptionSterile Plastic Pasteur Pipettes, 230 mm, Individually Wrap; 200/Case