Cole-Parmer Single Bracket for Filtration Cartridges with Fittings
Representative image only.
Cole-Parmer Single Bracket for Filtration Cartridges with Fittings
Cole-Parmer | Item: 0150672
Keep your filtration cartridges secure
₹90,200.00INR / Each
₹90,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Single Bracket for Filtration Cartridges with Fittings
Cole-Parmer Single Bracket for Filtration Cartridges with Fittings
0150672
₹90,200.00INR / Each
+ +
Cole-Parmer Ion Exchange Filter Cartridge, research
0150635
₹34,800.00INR / Each
+ =
Total Price 192400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Mount Cole-Parmer® Ion-X-changer and ResinTech IX series water purification cartridges
  • 18 month warranty
More About this Item
Choose from a variety mounting brackets to secure your filtration cartridges.
18 Month Warranty
Specifications
  • Compatible WithCole-Parmer® Ion-X-changer and ResinTech IX series water purification cartridges
  • DescriptionSingle Bracket for Filtration Cartridges with Fittings; 1/ea
  • Warranty18 months