Cole-Parmer® Ion-X-Changer Water Purification Cartridges
Purify water up to 15 MΩ-cm
  • Design your system with any combination of four different cartridges
Our four grades of disposable Ion-X-Changer filter cartridges withstand water pressures as high as 100 psi for safe hookup to any laboratory tap water system. Cartridge resins turn amber to indicate when cartridges need to be changed. All cartridges must be used with mounting brackets (order separately). Store processed water using the accumulator cartridge for faster dispensing.
Suggested System Combinations
A) One Adsorber and one Universal cartridge combination produces water free of all ionized constituents except free CO2 and silica.
B) Two Universal and two Research cartridges operating in tandem increase deionization capacity by 15%. Recommended for use with water that has a mineral salt concentration of 10 grains per gallon or more. When cartridges are mounted in pairs, use until the left cartridge is spent. Then, shift the right cartridge to the left side and replace the right side with a new cartridge. High water quality is retained.
C) Metex removes ≥99% of metallic ion, ammonia, and other cations. useful for power plants and for recovery of precious metals.
56,400.00 - ₹62,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Capacity (grains)
Availability Pricing
COMPARE
Universal Ion-X-Changer Water Purification Cartridge; 1/ea 1600 as Calcium Carbonate
₹62,100.00INR / Each
COMPARE
Adsorber Ion-X-Changer Water Purification Cartridge; 1/ea
₹59,700.00INR / Each
COMPARE
Research Ion-X-Changer Water Purification Cartridge; 1/ea 1000 as Calcium Carbonate
₹56,400.00INR / Each