Cole-Parmer SamplePrep 6807 Multivial Holder for Small and Microvials, 4 Vial Capacity
Representative image only.
Cole-Parmer SamplePrep 6807 Multivial Holder for Small and Microvials, 4 Vial Capacity
Cole-Parmer SamplePrep | Mfr: 6807| Item: 0450051
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
₹36,100.00INR / Each
₹36,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer SamplePrep 6807 Multivial Holder for Small and Microvials, 4 Vial Capacity
Cole-Parmer SamplePrep 6807 Multivial Holder for Small and Microvials, 4 Vial Capacity
0450051
₹36,100.00INR / Each
+ + + =
Total Price 359100.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Holds up to four small grinding vials and four microvial sets
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
This vial holder accommodates up to four small grinding vial sets or four microvial sets. This is ideal for grinding multiple samples in model CG-400, CG-450, or CG-500 Freezer/Mill® cryogenic grinders.
Specifications
  • Capacity4 small or microvials
  • DescriptionMultivial Holder for Small and Microvials, 4 Vial Capacity