IC Biomedical D050-8C00 Liquid Nitrogen Withdrawal System for Dewars 25 L and Larger
Representative image only.
IC Biomedical D050-8C00 Liquid Nitrogen Withdrawal System for Dewars 25 L and Larger
IC Biomedical | Mfr: D050-8C03| Item: 0377367
Permits safe and easy filling, venting, and transferring of liquid nitrogen
₹176,000.00INR / Each
₹176,000.00INR / Each
Buy It Together
Current Item
IC Biomedical D050-8C00 Liquid Nitrogen Withdrawal System for Dewars 25 L and Larger
IC Biomedical D050-8C00 Liquid Nitrogen Withdrawal System for Dewars 25 L and Larger
0377367
₹176,000.00INR / Each
+ =
Total Price 260000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Dispense liquid nitrogen at the rate of 8 L/min at 10 psi
  • 2 year warranty
More About this Item
Dispenses liquid nitrogen at the rate of 8 L/min at 10 psi. Unique design lets you fill the storage dewar without removing the withdrawal system or venting system pressure. For storage dewars 03773-57, -59, and -61.
The liquid withdrawal device enables the LD25, LD35, or LD50 dewars to conveniently pressure transfer liquid nitrogen at rates up to 8 liters per minute. The low pressure assures safe dispensing through a phase separator or an accessory transfer hose.
The liquid withdrawal device enables the LD25, LD35, or LD50 Dewars to conveniently pressure transfer liquid nitrogen at rates up to 8 liters per minute. The low pressure assures safe dispensing through a phase separator or an accessory transfer hose.
Two year warranty
Specifications
  • DescriptionLiquid Nitrogen Withdrawal System for Dewars 25 L and Larger