Cole-Parmer SamplePrep 6755 Rack for Small Grinding Vials, 16 Vial Capacity
Representative image only.
Cole-Parmer SamplePrep 6755 Rack for Small Grinding Vials, 16 Vial Capacity
Cole-Parmer SamplePrep | Mfr: 6755| Item: 0457632
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
₹7,900.00INR / Each
₹7,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer SamplePrep 6755 Rack for Small Grinding Vials, 16 Vial Capacity
Cole-Parmer SamplePrep 6755 Rack for Small Grinding Vials, 16 Vial Capacity
0457632
₹7,900.00INR / Each
+ + =
Total Price 183900.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Durable glass-reinforced acetal construction
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Holds up to sixteen of the following small grinding vials 04576-34, 04576-35, and 61043-07.
Specifications
  • Capacity16 small vials
  • DescriptionRack for Small Grinding Vials, 16 Vial Capacity