Cole-Parmer SamplePrep 2253C-48 Vial Cap for 50 mL Vial, Polyethylene; 48/PK
Representative image only.
Cole-Parmer SamplePrep 2253C-48 Vial Cap for 50 mL Vial, Polyethylene; 48/PK
Cole-Parmer SamplePrep | Mfr: 2253C-48| Item: 0460687
Safely grind samples in the HG-600 Geno/Grinder® 2010 and HG-400 MiniG® homogenizers
Call For Price
Call For Price
Buy It Together
Current Item
Cole-Parmer SamplePrep 2253C-48 Vial Cap for 50 mL Vial, Polyethylene; 48/PK
Cole-Parmer SamplePrep 2253C-48 Vial Cap for 50 mL Vial, Polyethylene; 48/PK
0460687
Call For Price
+ =
Total Price 41200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Screw thread cap ensures a tight seal
  • Compatible with 50 mL vial 04500-38
More About this Item
These polypropylene caps are lined with a thick 3/32” red silicone disc that prevents cracking. Threaded caps firmly seal Cole-Parmer® homogenizer vials. Replacement caps for 04576-99 and 04575-71. Vials sold separately.
Specifications
  • MaterialPolyethylene
  • ColorBlue
  • Compatible With04500-38 Vials
  • ConnectionsThread
  • DescriptionVial Cap for 50 mL Vial, Polyethylene; 48/PK