Cole-Parmer SamplePrep 2251C Vial Cap for 15 mL Short Vial, Polyethylene; 100/PK
Representative image only.
Cole-Parmer SamplePrep 2251C Vial Cap for 15 mL Short Vial, Polyethylene; 100/PK
Cole-Parmer SamplePrep | Mfr: 2251C| Item: 0460686
Safely grind samples in the HG-600 Geno/Grinder® 2010 and HG-400 MiniG® homogenizers
Call For Price
Call For Price
Buy It Together
Current Item
Cole-Parmer SamplePrep 2251C Vial Cap for 15 mL Short Vial, Polyethylene; 100/PK
Cole-Parmer SamplePrep 2251C Vial Cap for 15 mL Short Vial, Polyethylene; 100/PK
0460686
Call For Price
+ =
Total Price 41200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Screw thread cap ensures a tight seal
  • Compatible with 15 mL short vial 04576-99
More About this Item
These polypropylene caps are lined with a thick 3/32” red silicone disc that prevents cracking. Threaded caps firmly seal Cole-Parmer® homogenizer vials. Replacement caps for 04576-99 and 04575-71. Vials sold separately.
Specifications
  • MaterialPolyethylene
  • ColorWhite
  • Compatible With04576-99 Vials
  • ConnectionsThread
  • DescriptionVial Cap for 15 mL Short Vial, Polyethylene; 100/PK