Cole-Parmer SamplePrep 2211-10 Sealing Mat for Round-Bottom 96-Well Microplate; 10/PK
Representative image only.
Cole-Parmer SamplePrep 2211-10 Sealing Mat for Round-Bottom 96-Well Microplate; 10/PK
Cole-Parmer SamplePrep | Mfr: 2211-10| Item: 0450034
Tightly seals microplate wells to prevents spills and well-to-well contamination
₹11,400.00INR / Pkg of 10
₹11,400.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer SamplePrep 2211-10 Sealing Mat for Round-Bottom 96-Well Microplate; 10/PK
Cole-Parmer SamplePrep 2211-10 Sealing Mat for Round-Bottom 96-Well Microplate; 10/PK
0450034
₹11,400.00INR / Pkg of 10
+ + =
Total Price 152600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 96-well microplates with round-bottom wells (04500-31and-32)
  • Pack of 10
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. 
The sealing mat is constructed of rugged silicone rubber and designed to fit the 96-well microplates with round wells 04500-31 and 04500-32. May be sterilized and reused.
Specifications
  • MaterialSilicone
  • Compatible WithMicroplates 04500-31 and 04500-32
  • DescriptionSealing Mat for Round-Bottom 96-Well Microplate; 10/PK