Cole-Parmer Replaceable Membrane ISE Probe; Nitrate
Representative image only.
Cole-Parmer Replaceable Membrane ISE Probe; Nitrate
Cole-Parmer | Item: 2750512
Double-junction electrodes easily connect to meter or controller with BNC connection
₹90,100.00INR / Each
₹90,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer Replaceable Membrane ISE Probe; Nitrate
Cole-Parmer Replaceable Membrane ISE Probe; Nitrate
2750512
₹90,100.00INR / Each
+ =
Total Price 114600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Epoxy body provides durability
  • Color-coded PVC membranes for easy identification
  • 6 month warranty
More About this Item
The electrode features a durable outer epoxy body, making it perfect for field applications. The PVC membranes are color coded for easier identification. These double-junction combination electrodes require no external reference. Electrodes are refillable with an outer silver-free reference fill.
6 Month Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeNitrate
  • Max Temperature (° C)50
  • DescriptionReplaceable Membrane ISE Probe; Nitrate
  • Warranty6 month warranty