Cole-Parmer Replaceable Membrane ISE Probe; Chloride
Representative image only.
Cole-Parmer Replaceable Membrane ISE Probe; Chloride
Cole-Parmer | Item: 2750612
Don't throw it away—replace worn membranes quickly
₹78,300.00INR / Each
₹78,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer Replaceable Membrane ISE Probe; Chloride
Cole-Parmer Replaceable Membrane ISE Probe; Chloride
2750612
₹78,300.00INR / Each
+ +
Cole-Parmer ISE Electrode; Ammonium
2707701
₹68,300.00INR / Each
+ + +
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
₹22,300.00INR / Each
+ + =
Total Price 580500.00 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Replace worn membranes quickly
  • Extend the useful life of your electrodes
  • 6 month warranty
More About this Item
Membrane replacement saves time and the expense of electrode replacement — ideal for continuous or frequent sample analysis. The durable outer epoxy body is ideal for field applications and the PVC membranes are color coded for easier identification. To sense a new ion, interchange membranes of the same type. Simply replace the worn membrane without removing any other component of the electrode. The replaceable membrane is not part of the liquid junction, so adding the new membrane will not affect it. The reference electrolyte won’t leak during membrane replacement. These double-junction combination electrodes require no external reference. Electrodes are refillable with an outer silver-free reference fill.
Electrodes with PVC Membranes include one sensing module (in place), reference fill solution, internal fill solution, BNC connector with 3-ft cable, and instruction manual.
Electrodes with Solid-State Membranes include one sensing module (in place), polishing strips, reference fill solution, BNC connector with 3-ft cable, and instruction manual.
6 Month Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeChloride
  • Max Temperature (° C)80
  • DescriptionReplaceable Membrane ISE Probe; Chloride
  • Warranty6 month warranty