Cole-Parmer Removable High pH Double-Junction pH Electrode
Representative image only.
Cole-Parmer Removable High pH Double-Junction pH Electrode
Cole-Parmer | Item: 2700920
Easily remove electrodes in seconds without removing the holder
₹39,100.00INR / Each
₹39,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer Removable High pH Double-Junction pH Electrode
Cole-Parmer Removable High pH Double-Junction pH Electrode
2700920
₹39,100.00INR / Each
+ + =
Total Price 71500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
These electrodes are ideal for applications involving high-sodium concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Use for measurements from 0 to 14 pH. Electrodes withstand temperatures from 23 to 230ºF (-5 to 110ºC).
Note: 100Ω RTD ATC elements conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ thermistor ATC elements is a standard
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Min pH (pH)0
  • Max pH (pH)14
  • Max Temperature (° C)110
  • ConnectorBNC
  • DescriptionRemovable High pH Double-Junction pH Electrode