Cole-Parmer Printer
Representative image only.
Cole-Parmer Printer
Cole-Parmer | Item: 0301299
Accessories for your Cole-Parmer melting point apparatus
₹90,300.00INR / Each
₹90,300.00INR / Each
Product Description
Includes
90 to 250 VAC power adapter.
Key Features
  • Use with Cole-Parmer® MP-400D Digital Melting Point Apparatus and CC-200 Colony Counter
  • 3 year warranty
  • Previous part number SMP30/1
More About this Item
This printer has a 5-pin mini-DIN connection with a RS-232 serial interface and a 90 to 250 VAC power adapter.
3 Year WarrantyUL Listed to Canadian safety standards.
Specifications
  • Compatible WithMelting point apparatus 03011-49 and colony counters 14212-04 and 14212-06
  • DescriptionPrinter for melting point apparatus 03011-49 and colony counters 14212-04 and 14212-06
  • Warranty3 Years