Cole-Parmer pH Electrode for Ultrapure Water
Representative image only.
Cole-Parmer pH Electrode for Ultrapure Water
Cole-Parmer | Item: 2700450
Quickly and easily measure the pH of ultra pure water
₹37,500.00INR / Each
₹37,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer pH Electrode for Ultrapure Water
Cole-Parmer pH Electrode for Ultrapure Water
2700450
₹37,500.00INR / Each
+
Cole-Parmer Flow Cell for Ultrapure Water, 316 SS
2700400
₹87,600.00INR / Each
=
Total Price 125100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Constructed of durable stainless steel
  • Suitable for use with ultra pure water
  • 1 year warranty
More About this Item
Electrode has 10-ft long cable with BNC pH connector and tinned end for solution ground.
1 Year Warranty
Specifications
  • Electrode Body Material316 SS
  • Temperature CompensationNo
  • Thread Size NPT(M) (in)3/4
  • ConnectorBNC
  • DescriptionpH Electrode for Ultrapure Water