Cole-Parmer Flow Cell for Ultrapure Water, 316 SS
Representative image only.
Cole-Parmer Flow Cell for Ultrapure Water, 316 SS
Cole-Parmer | Item: 2700400
For in-line pH measurements of ultrapure water
₹87,600.00INR / Each
₹87,600.00INR / Each
Product Description
Key Features
  • Ideal for use in power plants
  • 1 year warranty
More About this Item
Measure the pH of high-purity water without the problems usually associated with low-ionic strength solutions. Concentric design and low sample flow rate (100 to 150 mL/min) help reduce streaming potential effects. Rugged 316 stainless steel system is grounded to prevent noise pickup and CO2 contamination. Electrode measures pH from 0 to 14 and withstands temperatures from 32 to 176°F (0 to 80°C). Order electrode, flow cell, and temperature compensation probe for a complete system. Electrode and temperature compensation probe sold separately.
Temperature probe (upper left), pH electrode (upper right), and flow cell (bottom) shown in image.
1 Year Warranty
Specifications
  • Electrode Body Material316 SS
  • Thread Size NPT(F) (in)1/4
  • DescriptionFlow Cell for Ultrapure Water, 316 SS