Cole-Parmer PFA Tubing, 5/32" ID x 1/4" OD; 50 Ft

Representative image only.

Cole-Parmer PFA Tubing, 5/32" ID x 1/4" OD; 50 Ft

Cole-Parmer | Item: 5011940

Superior micro-porosity characteristics and externally smooth finishes—ideal for high-purity applications

₹45,400.00INR / Pkg of 1

₹45,400.00INR / Pkg of 1

Buy It Together

Current Item

Cole-Parmer PFA Tubing, 5/32" ID x 1/4" OD; 50 Ft
Cole-Parmer PFA Tubing, 5/32" ID x 1/4" OD; 50 Ft
5011940

₹45,400.00INR / Pkg of 1
+ + + + + =
Total Price 90400.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Made from 100% virgin ultra-high molecular weight perfluoroalkoxy
  • Excellent flexibility compared to other PFA grade tubing
  • Wide temperature range of –160 to 260 °C (–320 to 500 °F)
  • Does not contain BPA, phthalates, or PVC
  • Sterilize by autoclave, EtO, and chemical sterilants
  • Complies with FDA 21 CFR 177.1550, USP Class VI, ASTM D 6867-03
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • 5/32" ID x 1/4" OD
  • Manufacturer defect warranty

More About this Item

The Cole-Parmer PFA tubing, made from ultra-high purity perfluoroalkoxy, is specifically designed to handle harsh chemicals and ultra-high purity applications. Smooth, nonstick surface finish and low permeability result in reduced absorption/adsorption of fluids, minimizing the risk of fluid contamination.

Great flexibility and resistance to cracks make PFA tubing perfect for use in moving parts. High thermal stability characteristics allow use up to 260 °C (500 °F). Translucent tubing makes it easy to view your fluid for sight gauge and flow applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Tubing FormulationPFA
  • Tubing OD (in)1/4
  • Tubing OD (mm)6.4
  • Tubing ID (in)5/32
  • Tubing ID (mm)4
  • Wall Thickness (in)3/64
  • Wall Thickness (mm)1.2
  • Clarity (translucent)Yes
  • Max Working Pressure at 73 F (PSI)352
  • Min Temperature (° F)-320
  • Max Temperature (° F)500
  • Min Temperature (° C)-160
  • Max Temperature (° C)260
  • Bend Radius (in)3/4
  • Bend Radius (mm)19
  • Length (ft)50
  • Sterilization MethodAutoclave;EtO
  • Regulatory ApprovalsFDA;RoHS;Non-Phthalate;ADCF;USP
  • Durometer60 Shore D
  • DescriptionPFA Tubing, 5/32" ID x 1/4" OD; 50 Ft
Documents & Certificates

Certificate of Compliance

Reviews
Q&A