Cole-Parmer Perforated Stainless Steel Insert Tray for 2.5 Gal Ultrasonic Cleaners
Representative image only.
Cole-Parmer Perforated Stainless Steel Insert Tray for 2.5 Gal Ultrasonic Cleaners
Cole-Parmer | Item: 0885342
Achieve optimal cleaning results
₹49,600.00INR / Each
₹49,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer Perforated Stainless Steel Insert Tray for 2.5  Gal Ultrasonic Cleaners
Cole-Parmer Perforated Stainless Steel Insert Tray for 2.5 Gal Ultrasonic Cleaners
0885342
₹49,600.00INR / Each
+ + + =
Total Price 416600.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Supports items that require direct cleaning
  • Prevents damage to transducers
  • 1 year warranty
More About this Item
Made of 304 stainless steel, these perforated trays can be used to support items in the tank that require direct cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners. Use an insert tray or beaker positioning cover and beaker with these ultrasonic cleaners. Ultrasonic acticity is greatest starting about an inch from the bottom of the tank.
1 Year Warranty
Specifications
  • Capacity (Gal)2.5
  • Capacity (Liters)9.46
  • Length (InchShort)11
  • Width (InchShort)9
  • Height (InchShort)6
  • Length (cm)27.94
  • Width (cm)22.86
  • Height (cm)15.24
  • Material304 Stainless Steel
  • DescriptionPerforated Stainless Steel Insert Tray for 2.5 Gal Ultrasonic Cleaners