Cole-Parmer® UC-200 Series Ultrasonic Cleaners Baskets
Provides optimal cleaning results
  • Supports items that require indirect cleaning
  • Legs suspend items safely away from bottom of tank
  • Convenient handles allow for safe and easy repositioning of objects
  • Mesh size opening: 10 x 10 mm
These wire mesh metal ultrasonic cleaner baskets are ideal for supporting materials, instruments, and parts that require indirect cleaning. Prevents damage by keeping items from touching bottom of the tank. Side handles allow you to easily reposition objects being cleaned during the ultrasonic cleaning cycle.
3,600.00 - ₹13,200.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Height (cm) Length (cm)
Availability Pricing
COMPARE
0.5 12.8 13.8
₹3,600.00INR / Each
COMPARE
0.75 12.5 22.9
₹4,000.00INR / Each
COMPARE
1.5 11.6 26.5
₹4,900.00INR / Each
COMPARE
2.5 20.4 26.4
₹5,500.00INR / Each
COMPARE
3.5 26.4 29.4
₹7,000.00INR / Each
COMPARE
5.25 26.3 45.8
₹9,100.00INR / Each
COMPARE
7.25 26.3 45.8
₹13,200.00INR / Each
Showing 1- 7 of 7