Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 500mL; 12/PK

Representative image only.

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 500mL; 12/PK

Cole-Parmer | Item: 0625340

Transport hazardous samples reliably

₹9,300.00INR / Pkg of 12

₹9,300.00INR / Pkg of 12

Buy It Together

Current Item

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 500mL; 12/PK
Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 500mL; 12/PK
0625340

₹9,300.00INR / Pkg of 12
+
Cole-Parmer Replacement Cap, PP, 28mm; 12/PK
6300006

₹2,900.00INR / Pkg of 12
=
Total Price 12200.00 ADD 2 ITEMS TO CART
Product Description

Includes

Polypropylene screw closures.

Key Features

  • Safe for small quantities of corrosives or cleaning supplies
  • Leakproof, PP screw cap
  • 1 year warranty

More About this Item

Ideal for small-scale inter-lab shipping of hazardous samples and chemicals such as corrosives or cleaning compounds.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

1 Year Warranty
Specifications
  • Capacity (mL)500
  • Capacity (L)0.5
  • Capacity (oz)16
  • Capacity (Gal)0.132100396301189
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene
  • Cap / Stopper Size28
  • Cap Colornatural
  • Bottle Colornatural
  • AutoclavableNo
  • Neck Inside Diameter (ID) (mm)19.6
  • Neck Inside Diameter (ID) (in)49/64
  • Outside Diameter (OD) (in)2 29/32
  • Outside Diameter (OD) (cm)7.4
  • Overall Height (in)6 11/16
  • Overall Height (cm)17
  • Overall Depth (in)2 29/32
  • Overall Depth (cm)7.4
  • Overall Width (in)2 29/32
  • Overall Width (cm)7.4
  • DescriptionNarrow-Mouth Transport Plastic Bottle, HDPE, 500mL; 12/PK
  • Warranty1 year
Reviews
Q&A