Cole-Parmer Microscope Slides, Hydrophilic Adhesive, White-Frosted, 45° Corners; 72/PK

Representative image only.

Cole-Parmer Microscope Slides, Hydrophilic Adhesive, White-Frosted, 45° Corners; 72/PK

Cole-Parmer | Item: 7890591

Specially prepared slides enhance adhesion of tissue samples to slides

₹2,600.00INR / Pkg of 72

₹2,600.00INR / Pkg of 72

Buy It Together

Current Item

Cole-Parmer Microscope Slides, Hydrophilic Adhesive, White-Frosted, 45° Corners; 72/PK
Cole-Parmer Microscope Slides, Hydrophilic Adhesive, White-Frosted, 45° Corners; 72/PK
7890591

₹2,600.00INR / Pkg of 72
+
Cole-Parmer Microscope Cover Glasses, 18 x 18 mm; 100/PK
7890593

₹320.00INR / Pkg of 100
=
Total Price 2920.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Frosted one end on one side
  • Ground edges and 45° corners reduce the chance of cutting your fingers or tearing of gloves
  • Each slide measures 76 x 26 mm and 1.0 to 1.2-mm thick
  • Hydrophilic adhesive slides offer excellent tissue adhesion and staining performance
  • 2 year warranty

More About this Item

Chemically resistant white writing surface is suitable for marking with inkjet and thermal transfer printers and permanent markers.

Hydrophilic adhesive slides carry more positive charges that keep tissue from falling off slides. Hydrophilic surface helps the water-based reagent to cover the sections evenly, which improve staining as well as reduce false negative and the background staining.

Specifications
  • Slide TypeHydrophilic adhesive
  • Thickness1.0 to 1.2 mm
  • Size76 x 26 mm
  • DescriptionMicroscope Slides, Hydrophilic Adhesive, White-Frosted, 45° Corners; 72/PK
  • Warranty2 Years
Reviews
Q&A