Cole-Parmer Micro-90 Cleaning Solution; 5 Gal Bottle
Representative image only.
Cole-Parmer Micro-90 Cleaning Solution; 5 Gal Bottle
Cole-Parmer | Item: 1810020
Effective Detergent for Glass, Metal, Plastic, and Rubber Surfaces
₹89,000.00INR / Each
₹89,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Micro-90 Cleaning Solution; 5 Gal Bottle
Cole-Parmer Micro-90 Cleaning Solution; 5 Gal Bottle
1810020
₹89,000.00INR / Each
+ + + + +
Texwipe TX3340 Cleaning Validation Kit, 12 Areas; 18/CS
3367640
₹652,000.00INR / Case of 18
+ + +
Small Bottle Brush Folding Tip 12", 12/pk
8455086
₹36,500.00INR / Pkg of 12
=
Total Price 849300.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Powerful Cleaning Performance: Micro-90 is a mild yet effective, free-rinsing cleaner suitable for both critical and general-purpose cleaning applications. It efficiently removes most biological materials, including blood and proteins, ensuring thorough decontamination.
  • Versatile Application: Safe for use on glass, most metals, plastics, rubber, and fabrics. Compatible with all ultrasonic cleaners, making it ideal for laboratories, industrial settings, and healthcare facilities.
  • Environmentally Friendly: The biodegradable, phosphate-free formula ensures minimal environmental impact, aligning with sustainable practices.
  • Flexible Usage: Effective at various concentrations, with most applications requiring only 1.25 to 2.5 fluid ounces per gallon of water.
More About this Item
Achieve superior cleaning performance with Cole-Parmer Micro-90 Biodegradable Cleaning Solution, a versatile, phosphate-free detergent designed for manual and ultrasonic cleaning. This high-performance, non-ionic cleaner effectively removes biological residues, oils, and contaminants from a wide range of surfaces, ensuring thorough decontamination in laboratories, industrial environments, and healthcare facilities.
Do not use for private purposes (household).
Specifications
  • FormatLiquid
  • Volume (gallons)5
  • Volume (Liters)20
  • Dilution1:100 to 2:100, 9.5 pH
  • ApplicationsManual or ultrasonic; removes blood and most biologoical materials; cleans glass, most metals, plastic, and rubber
  • DescriptionMicro-90 cleaning solution, 5 gallon bottle