Cole-Parmer Mesh Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
Representative image only.
Cole-Parmer Mesh Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
Cole-Parmer | Item: 0885045
Achieve optimal cleaning results
₹25,500.00INR / Each
₹25,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer Mesh Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
Cole-Parmer Mesh Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
0885045
₹25,500.00INR / Each
+ + + =
Total Price 393400.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Supports items in tank for direct cleaning
  • Prevents damage to transducers
More About this Item
Made of 304 stainless steel, these mesh trays can be used to support items in the tank that require direct cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers. Features #4 mesh with .25 inch openings.
Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners. Use an insert tray or beaker positioning cover and beaker with these ultrasonic cleaners. Ultrasonic acticity is greatest starting about an inch from the bottom of the tank.
Specifications
  • Capacity (Gal)0.5
  • Capacity (Liters)1.89
  • Length (InchShort)5
  • Width (InchShort)4 1/2
  • Height (InchShort)3
  • Length (cm)12.7
  • Width (cm)11.43
  • Height (cm)7.62
  • DescriptionMesh Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners