Cole-Parmer Melting Point Calibration Kit
Representative image only.
Cole-Parmer Melting Point Calibration Kit
Cole-Parmer | Item: 9996749
Accessories for your Cole-Parmer melting point apparatus
Call For Price
Call For Price
Product Description
Key Features
  • Ensure the accuracy of your melting point apparatus
  • 1 year warranty
More About this Item
Comes with three WHO approved melting point standards—caffeine, vanillin, and sulphanilamide—with purity certificates.
1 Year Warranty
Specifications
  • Compatible With03013-02 and 03013-03
  • DescriptionMelting Point Calibration Kit